Learn More
Alkali Scientific p Nitrophenyl β D fucopyranoside, 1 G
SDP
Catalog No. 5000660282
Encompass_Preferred
p Nitrophenyl β-D fucopyranoside (1 g) is a high-purity reagent designed for use in enzymatic assays where fucosidases are involved. This reagent is commonly utilized in biochemical studies to detect and measure the activity of β-D-fucosidases through colorimetric analysis. Upon hydrolysis, the nitrophenyl group releases a yellow color, enabling precise measurement of enzyme activity. The compound is ideal for high-throughput assays, carbohydrate research, and studies involving glycoside hydrolases. It plays a key role in understanding the breakdown of fucosidic linkages in glycoproteins, a process important in cellular communication and immune response.
Catalog No. 50-006-60282 Supplier Alkali Scientific Supplier No. BDF003CP
p Nitrophenyl β-D fucopyranoside (1 g) is a high-purity reagent designed for use in enzymatic assays where fucosidases are involved. This reagent is commonly utilized in biochemical studies to detect and measure the activity of β-D-fucosidases through colorimetric analysis. Upon hydrolysis, the nitrophenyl group releases a yellow color, enabling precise measurement of enzyme activity. The compound is ideal for high-throughput assays, carbohydrate research, and studies involving glycoside hydrolases. It plays a key role in understanding the breakdown of fucosidic linkages in glycoproteins, a process important in cellular communication and immune response.