Learn More
Alkali Scientific p Nitrophenyl β D fucopyranoside, 100 Mg
SDP
Catalog No. 5000659859
Encompass_Preferred
p Nitrophenyl β-D fucopyranoside is a biochemical reagent used in carbohydrate and enzyme studies, particularly in glycoside hydrolase assays. This 100 mg reagent is specifically designed for use in experiments that require a chromogenic substrate for β-D-fucosidases. Upon hydrolysis by the appropriate enzyme, the nitrophenyl group releases a yellow color, making it easy to measure enzyme activity. This reagent is often used in the study of glycosylation patterns and enzymatic catalysis related to fucose-containing glycoproteins. It is suitable for high-throughput screening and biochemical analysis in various research fields, including molecular biology and carbohydrate chemistry.
Catalog No. 50-006-59859 Supplier Alkali Scientific Supplier No. BDF001CP
p Nitrophenyl β-D fucopyranoside is a biochemical reagent used in carbohydrate and enzyme studies, particularly in glycoside hydrolase assays. This 100 mg reagent is specifically designed for use in experiments that require a chromogenic substrate for β-D-fucosidases. Upon hydrolysis by the appropriate enzyme, the nitrophenyl group releases a yellow color, making it easy to measure enzyme activity. This reagent is often used in the study of glycosylation patterns and enzymatic catalysis related to fucose-containing glycoproteins. It is suitable for high-throughput screening and biochemical analysis in various research fields, including molecular biology and carbohydrate chemistry.