Learn More
Alkali Scientific p Nitrophenyl β D fucopyranoside, 250 Mg
SDP
Catalog No. 5000660671
Encompass_Preferred
p Nitrophenyl β-D fucopyranoside (250 mg) is a highly effective substrate for studying fucosidase activity in biochemical assays. This reagent is commonly used in research related to carbohydrate enzymes and glycosidases, particularly in the breakdown of fucosidic linkages in glycoproteins and glycolipids. The compound releases a yellow color upon enzymatic hydrolysis, making it ideal for colorimetric analysis. Its application is especially useful in assays that require the measurement of fucosidase activity, aiding in research focused on glycosylation, enzyme kinetics, and the role of fucose in biological systems such as cell signaling and immunity.
Catalog No. 50-006-60671 Supplier Alkali Scientific Supplier No. BDF002CP
p Nitrophenyl β-D fucopyranoside (250 mg) is a highly effective substrate for studying fucosidase activity in biochemical assays. This reagent is commonly used in research related to carbohydrate enzymes and glycosidases, particularly in the breakdown of fucosidic linkages in glycoproteins and glycolipids. The compound releases a yellow color upon enzymatic hydrolysis, making it ideal for colorimetric analysis. Its application is especially useful in assays that require the measurement of fucosidase activity, aiding in research focused on glycosylation, enzyme kinetics, and the role of fucose in biological systems such as cell signaling and immunity.