Learn More
Alkali Scientific p Nitrophenyl β D fucopyranoside, 5 G
SDP
Catalog No. 5000661160
Encompass_Preferred
p Nitrophenyl β-D fucopyranoside (5 g) is a versatile biochemical reagent used in the study of fucosidase enzymes. This substrate is particularly useful in assays that measure the activity of β-D-fucosidases, enzymes that hydrolyze fucosidic bonds in glycoproteins and glycolipids. The hydrolysis process releases a yellow color, allowing for easy detection and quantification of enzymatic activity. This reagent is frequently used in research on carbohydrate metabolism, glycosylation patterns, and enzyme kinetics, as well as in high-throughput screening for identifying new fucosidases or inhibitors in drug discovery and molecular biology.
Catalog No. 50-006-61160 Supplier Alkali Scientific Supplier No. BDF004CP
p Nitrophenyl β-D fucopyranoside (5 g) is a versatile biochemical reagent used in the study of fucosidase enzymes. This substrate is particularly useful in assays that measure the activity of β-D-fucosidases, enzymes that hydrolyze fucosidic bonds in glycoproteins and glycolipids. The hydrolysis process releases a yellow color, allowing for easy detection and quantification of enzymatic activity. This reagent is frequently used in research on carbohydrate metabolism, glycosylation patterns, and enzyme kinetics, as well as in high-throughput screening for identifying new fucosidases or inhibitors in drug discovery and molecular biology.