Learn More
Alkali Scientific p Nitrophenyl β D Galactopyranoside, 5 G
SDP
Catalog No. 5000660436
Encompass_Preferred
p-Nitrophenyl β-D Galactopyranoside is a chemical reagent commonly used in biochemical assays to study enzyme activity, particularly in research involving β-galactosidase. This compound is highly useful in the detection and quantification of β-galactosidase, an enzyme often employed in genetic research and diagnostics. The reagent is utilized in colorimetric assays, where the cleavage of the β-galactoside bond releases a yellow-colored product, indicating enzyme activity. This 5g package provides a convenient quantity for laboratory applications and is widely used in enzyme assays, microbiological studies, and genetic research. It is a highly reliable and effective tool for routine and experimental biochemical analysis in research laboratories.
Catalog No. 50-006-60436 Supplier Alkali Scientific Supplier No. TOP002CP
p-Nitrophenyl β-D Galactopyranoside is a chemical reagent commonly used in biochemical assays to study enzyme activity, particularly in research involving β-galactosidase. This compound is highly useful in the detection and quantification of β-galactosidase, an enzyme often employed in genetic research and diagnostics. The reagent is utilized in colorimetric assays, where the cleavage of the β-galactoside bond releases a yellow-colored product, indicating enzyme activity. This 5g package provides a convenient quantity for laboratory applications and is widely used in enzyme assays, microbiological studies, and genetic research. It is a highly reliable and effective tool for routine and experimental biochemical analysis in research laboratories.