Learn More
Alkali Scientific p Nitrophenyl β D Galactopyranoside, 50 G
SDP
Catalog No. 5000661052
Encompass_Preferred
p-Nitrophenyl β-D Galactopyranoside, 50g, is a highly concentrated reagent used for studying β-galactosidase activity in biochemical assays. This compound serves as a substrate in enzymatic reactions where β-galactosidase cleaves the β-galactoside bond to release a yellow-colored product. This colorimetric assay is commonly employed in molecular biology, microbiology, and gene expression studies to monitor enzyme activity. It is an essential reagent for researchers working with bacterial cultures, genetic modifications, and enzymatic processes, offering an easy-to-use and reliable method for enzyme analysis. The 50g quantity ensures ample material for large-scale experiments and routine laboratory procedures.
Catalog No. 50-006-61052 Supplier Alkali Scientific Supplier No. TOP003CP
p-Nitrophenyl β-D Galactopyranoside, 50g, is a highly concentrated reagent used for studying β-galactosidase activity in biochemical assays. This compound serves as a substrate in enzymatic reactions where β-galactosidase cleaves the β-galactoside bond to release a yellow-colored product. This colorimetric assay is commonly employed in molecular biology, microbiology, and gene expression studies to monitor enzyme activity. It is an essential reagent for researchers working with bacterial cultures, genetic modifications, and enzymatic processes, offering an easy-to-use and reliable method for enzyme analysis. The 50g quantity ensures ample material for large-scale experiments and routine laboratory procedures.