Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Nitrophenyl-α-D-galactopyranoside, 98%, MP Biomedicals™

Catalog No. ICN10247101 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10247101 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10247101 Supplier MP Biomedicals, Inc Supplier No. 0210247101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7493-95-0 | C12H15NO8 | 301.251 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7493-95-0
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.251
InChI Key IFBHRQDFSNCLOZ-IIRVCBMXSA-N
Synonym 4-nitrophenyl a-d-galactopyranoside, 4-nitrophenyl-alpha-d-galactopyranoside, pnpalphagal, p-nitrophenyl-alpha-d-galactopyranoside, 4-nitrophenyl, a-d-galactopyranoside, p-nitrophenyl alpha-d-galactopyranoside, 4-nitrophenyl alpha-d-galactopyranoside, p-nitrophenyl-alpha-d-galactoside, 4-nitrophenyl-alpha-d-galactoside, 1-o-4-nitrophenyl-alpha-d-galactose
PubChem CID 82000
ChEBI CHEBI:546840
IUPAC Name (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O

Specifications

Color White
Assay Percent Range 0.98
Quantity 1 g
Formula Weight 301.3
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material p-Nitrophenyl-α-D-galactopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.