Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Nitrophenyl-β-D-galactopyranoside, >99.75%, MP Biomedicals™

Catalog No. icn10247205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10247205 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10247205 Supplier MP Biomedicals, Inc Supplier No. 0210247205
Only null left
Add to Cart
Add to Cart

Specifications

CAS 3150-24-1
Color White
Assay Percent Range > 99.75%
Molecular Formula C12H15NO8
Quantity 5 g
MDL Number MFCD00063256
Synonym 4-nitrophenyl-beta-d-galactopyranoside, 4-nitrophenylgalactoside, p-nitrophenyl-beta-d-galactoside, p-nitrophenyl beta-d-galactopyranoside, 4-nitrophenyl beta-d-galactopyranoside, p-nitrophenol galactoside, 4-nitrophenyl-beta-d-galactoside, 4-nitrophenyl galactoside, 1-o-p-nitrophenyl-beta-d-galactopyranose, 4-nitrophenylbeta-d-galactopyranoside
InChI Key IFBHRQDFSNCLOZ-YBXAARCKSA-N
SMILES OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular Weight (g/mol) 301.25
PubChem CID 65115
ChEBI CHEBI:355715
Formula Weight 301.3
Physical Form Crystalline
Chemical Name or Material p-Nitrophenyl-β-D-galactopyranoside, For TLC analysis
Show More Show Less
EINECSNumber 221-584-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.