Learn More
MP Biomedicals™ P-Nitrophenyl-α-D-Galactopyranoside, MP Biomedicals™
Shop All MP Biomedicals, Inc ProductsSpecifications
Specifications
| Form | Powder |
| Quantity | 100mg |
| Format | White Powder |
| Molecular Weight (g/mol) | 301.3 |
| Storage Requirements | 0°C, protect from light |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.