Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ P-Nitrophenyl-α-D-Galactopyranoside, MP Biomedicals™

Catalog No. MP021024715 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021024715 500mg
MP021024712 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021024715 Supplier MP Biomedicals™ Supplier No. 02102471.5
Add to Cart
Add to Cart

P-Nitrophenyl-α-D-Galactopyranoside, MP Biomedicals is used as a substrate for α-galactosidase.

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Quantity 500mg
Format White Powder
Molecular Weight (g/mol) 301.3
Storage Requirements 0°C, protect from light
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.