Learn More
Alkali Scientific p Nitrophenyl α D glucopyranoside [4 Nitrophenyl α D glucoside], 1 G
SDP
Catalog No. 5000661893
Encompass_Preferred
p Nitrophenyl α-D glucopyranoside (1 g) is a commonly used biochemical reagent for studying α-D-glucosidases in enzyme assays. This compound releases a yellow color when hydrolyzed by α-D-glucosidases, making it ideal for colorimetric assays. The reagent is essential in glycobiology for studying the enzymatic breakdown of glucosidic linkages in various substrates. Its use spans across research on carbohydrate metabolism, enzyme kinetics, and the development of new enzymes or inhibitors. The compound is particularly valuable for high-throughput screening and various applications in carbohydrate chemistry, including the analysis of glucosidases involved in cellular processes.
Catalog No. 50-006-61893 Supplier Alkali Scientific Supplier No. RAN001CP
p Nitrophenyl α-D glucopyranoside (1 g) is a commonly used biochemical reagent for studying α-D-glucosidases in enzyme assays. This compound releases a yellow color when hydrolyzed by α-D-glucosidases, making it ideal for colorimetric assays. The reagent is essential in glycobiology for studying the enzymatic breakdown of glucosidic linkages in various substrates. Its use spans across research on carbohydrate metabolism, enzyme kinetics, and the development of new enzymes or inhibitors. The compound is particularly valuable for high-throughput screening and various applications in carbohydrate chemistry, including the analysis of glucosidases involved in cellular processes.