Learn More
Alkali Scientific p Nitrophenyl α D glucopyranoside [4 Nitrophenyl α D glucoside], 50 Grams
SDP
Catalog No. 5000661699
Encompass_Preferred
Nitrophenyl alpha-D-glucopyranoside (50 g) is a chromogenic substrate used in enzyme assays to detect glucosidase activity. The hydrolysis of this substrate by glucosidase enzymes releases a yellow product, allowing easy visual or quantitative detection of enzyme activity. This reagent is commonly used in biochemical research, including enzyme kinetics, metabolic studies, and the study of carbohydrate hydrolysis. The 50 g formulation is ideal for large-scale applications in microbiology, diagnostics, and pharmaceutical research. It is particularly useful in identifying microorganisms that produce glucosidases and in studying enzyme activity in various biological samples.
Catalog No. 50-006-61699 Supplier Alkali Scientific Supplier No. RAN004CP
Nitrophenyl alpha-D-glucopyranoside (50 g) is a chromogenic substrate used in enzyme assays to detect glucosidase activity. The hydrolysis of this substrate by glucosidase enzymes releases a yellow product, allowing easy visual or quantitative detection of enzyme activity. This reagent is commonly used in biochemical research, including enzyme kinetics, metabolic studies, and the study of carbohydrate hydrolysis. The 50 g formulation is ideal for large-scale applications in microbiology, diagnostics, and pharmaceutical research. It is particularly useful in identifying microorganisms that produce glucosidases and in studying enzyme activity in various biological samples.