Learn More
Alkali Scientific p Nitrophenyl β D mannopyranoside, 1 G
SDP
Catalog No. 5000660203
Encompass_Preferred
Nitrophenyl β-D-mannopyranoside is a chemical reagent provided in a 1g quantity, used primarily for measuring the activity of β-mannosidase in biochemical and enzymatic studies. This substrate is essential for understanding the hydrolysis of mannosidic bonds in mannan and other polysaccharides. It plays an important role in the study of carbohydrate metabolism, enzymatic breakdown of plant polysaccharides, and the enzymatic processing of sugars in microbes. This reagent is valuable in research areas such as glycobiology, proteomics, and enzymology, offering insights into enzyme kinetics, carbohydrate degradation, and the biological function of β-mannosidase.
Catalog No. 50-006-60203 Supplier Alkali Scientific Supplier No. BDM003CP
Nitrophenyl β-D-mannopyranoside is a chemical reagent provided in a 1g quantity, used primarily for measuring the activity of β-mannosidase in biochemical and enzymatic studies. This substrate is essential for understanding the hydrolysis of mannosidic bonds in mannan and other polysaccharides. It plays an important role in the study of carbohydrate metabolism, enzymatic breakdown of plant polysaccharides, and the enzymatic processing of sugars in microbes. This reagent is valuable in research areas such as glycobiology, proteomics, and enzymology, offering insights into enzyme kinetics, carbohydrate degradation, and the biological function of β-mannosidase.