Learn More
Medchemexpress LLC P-nitrophenyl α-D-mannopyranoside | 10357-27-4 | 98.88% | 301.25 | 250 MG
SDP
Catalog No. 5000348785
Encompass_Preferred
P-nitrophenyl α-D-mannopyranoside is a chromogenic glycosidic ligand. This compound features a sugar moiety (α-D-mannopyranose) with a mannose-characteristic axial hydroxyl group at the C2 position, and a ligand portion (p-nitrophenol) that provides ultraviolet-visible light absorption properties. It is used to determine glycosidase activity and to characterize the sugar recognition properties of lectins.
  • Chromogenic glycosidic ligand
  • Features an α-D-mannopyranose sugar moiety
  • Contains a p-nitrophenol ligand portion
  • Provides ultraviolet-visible light absorption properties
  • Suitable for determining glycosidase activity
  • Used to characterize lectin sugar recognition properties
Catalog No. 50-003-48785 Supplier Medchemexpress LLC Supplier No. 250MGHY137822
P-nitrophenyl α-D-mannopyranoside is a chromogenic glycosidic ligand. This compound features a sugar moiety (α-D-mannopyranose) with a mannose-characteristic axial hydroxyl group at the C2 position, and a ligand portion (p-nitrophenol) that provides ultraviolet-visible light absorption properties. It is used to determine glycosidase activity and to characterize the sugar recognition properties of lectins.
  • Chromogenic glycosidic ligand
  • Features an α-D-mannopyranose sugar moiety
  • Contains a p-nitrophenol ligand portion
  • Provides ultraviolet-visible light absorption properties
  • Suitable for determining glycosidase activity
  • Used to characterize lectin sugar recognition properties