Learn More
Alkali Scientific p Nitrophenyl α D mannopyranoside, 25 G
SDP
Catalog No. 5000659838
Encompass_Preferred
p-Nitrophenyl α-D Mannopyranoside, 25g, is a highly concentrated reagent used for studying α-mannosidase activity. In enzymatic assays, α-mannosidase cleaves the β-galactosidase bond in the substrate, releasing a yellow product, which serves as a visual indicator of enzyme activity. This reagent is widely used in enzyme kinetics studies, particularly in microbiology, genetic research, and carbohydrate metabolism. The 25g size is ideal for large-scale experiments or routine laboratory applications. It is particularly valuable for researchers who frequently need to measure α-mannosidase activity in a variety of samples, offering a cost-effective and efficient reagent for enzymatic analysis.
Catalog No. 50-006-59838 Supplier Alkali Scientific Supplier No. LAD003CP
Add to Cart
Add to Cart
p-Nitrophenyl α-D Mannopyranoside, 25g, is a highly concentrated reagent used for studying α-mannosidase activity. In enzymatic assays, α-mannosidase cleaves the β-galactosidase bond in the substrate, releasing a yellow product, which serves as a visual indicator of enzyme activity. This reagent is widely used in enzyme kinetics studies, particularly in microbiology, genetic research, and carbohydrate metabolism. The 25g size is ideal for large-scale experiments or routine laboratory applications. It is particularly valuable for researchers who frequently need to measure α-mannosidase activity in a variety of samples, offering a cost-effective and efficient reagent for enzymatic analysis.