Learn More
Alkali Scientific p Nitrophenyl β D mannopyranoside, 250 Mg
SDP
Catalog No. 5000660202
Encompass_Preferred
Nitrophenyl β-D-mannopyranoside is a chemical reagent provided in a 250mg quantity for enzymatic assays and biochemical research. This substrate is commonly used to test the activity of β-mannosidase, an enzyme involved in the breakdown of mannan and other related polysaccharides. The reagent is valuable for studying carbohydrate metabolism, glycoside hydrolases, and enzyme specificity. By using this reagent, researchers can investigate the catalytic activity of β-mannosidase in hydrolyzing β-mannosidic linkages, which plays an important role in plant cell wall degradation, microbial metabolism, and enzymatic reactions in both natural and industrial processes.
Catalog No. 50-006-60202 Supplier Alkali Scientific Supplier No. BDM002CP
Nitrophenyl β-D-mannopyranoside is a chemical reagent provided in a 250mg quantity for enzymatic assays and biochemical research. This substrate is commonly used to test the activity of β-mannosidase, an enzyme involved in the breakdown of mannan and other related polysaccharides. The reagent is valuable for studying carbohydrate metabolism, glycoside hydrolases, and enzyme specificity. By using this reagent, researchers can investigate the catalytic activity of β-mannosidase in hydrolyzing β-mannosidic linkages, which plays an important role in plant cell wall degradation, microbial metabolism, and enzymatic reactions in both natural and industrial processes.