Learn More
Alkali Scientific p Nitrophenyl α L Arabinofuranoside, 100 Mg
SDP
Catalog No. 5000660821
Encompass_Preferred
Nitrophenyl α-L-arabinofuranoside is a reagent supplied in a 100mg quantity for biochemical research and enzymatic assays. This substrate is specifically used to test the activity of enzymes like α-arabinofuranosidase, which hydrolyze arabinofuranoside bonds. The reagent serves as an effective tool in the study of carbohydrate metabolism, particularly in research on hemicellulose and pectin degradation. It is widely used in glycosidic bond cleavage studies to understand enzymatic behavior and substrate specificity. This reagent is suitable for applications in plant biochemistry, microbiology, and environmental science, where arabinofuranoside is a key component.
Catalog No. 50-006-60821 Supplier Alkali Scientific Supplier No. ALA002CP
Nitrophenyl α-L-arabinofuranoside is a reagent supplied in a 100mg quantity for biochemical research and enzymatic assays. This substrate is specifically used to test the activity of enzymes like α-arabinofuranosidase, which hydrolyze arabinofuranoside bonds. The reagent serves as an effective tool in the study of carbohydrate metabolism, particularly in research on hemicellulose and pectin degradation. It is widely used in glycosidic bond cleavage studies to understand enzymatic behavior and substrate specificity. This reagent is suitable for applications in plant biochemistry, microbiology, and environmental science, where arabinofuranoside is a key component.