Learn More
Alkali Scientific p Nitrophenyl α L Arabinofuranoside, 25 Mg
SDP
Catalog No. 5000660816
Encompass_Preferred
Nitrophenyl α-L-arabinofuranoside is a chemical reagent provided in a 25mg quantity for enzymatic and biochemical applications. This substrate is commonly used in enzyme activity assays to study the hydrolysis of arabinofuranoside bonds. It is particularly useful for testing the activity of α-arabinofuranosidase, an enzyme involved in the degradation of hemicellulose and pectin. This reagent is ideal for research in plant biology, enzymology, and carbohydrate metabolism. By using this compound, researchers can examine the enzymatic breakdown of polysaccharides and explore their biological functions in various organisms, including plants and microorganisms.
Catalog No. 50-006-60816 Supplier Alkali Scientific Supplier No. ALA001CP
Nitrophenyl α-L-arabinofuranoside is a chemical reagent provided in a 25mg quantity for enzymatic and biochemical applications. This substrate is commonly used in enzyme activity assays to study the hydrolysis of arabinofuranoside bonds. It is particularly useful for testing the activity of α-arabinofuranosidase, an enzyme involved in the degradation of hemicellulose and pectin. This reagent is ideal for research in plant biology, enzymology, and carbohydrate metabolism. By using this compound, researchers can examine the enzymatic breakdown of polysaccharides and explore their biological functions in various organisms, including plants and microorganisms.