Learn More
Alkali Scientific p Nitrophenyl α L Arabinofuranoside, 500 Mg
SDP
Catalog No. 5000660822
Encompass_Preferred
Nitrophenyl α-L-arabinofuranoside is a chemical reagent available in a 500mg quantity, designed for use in biochemical assays to measure enzyme activity. This compound is a substrate for α-arabinofuranosidase, an enzyme involved in the breakdown of arabinose-containing polysaccharides. The reagent plays a critical role in studying the enzymatic hydrolysis of arabinofuranoside bonds, particularly in carbohydrate research and the degradation of plant cell wall components. It is widely used in enzymology, glycobiology, and proteomics research to explore the mechanisms of glycoside hydrolases and their role in carbohydrate metabolism.
Catalog No. 50-006-60822 Supplier Alkali Scientific Supplier No. ALA003CP
Nitrophenyl α-L-arabinofuranoside is a chemical reagent available in a 500mg quantity, designed for use in biochemical assays to measure enzyme activity. This compound is a substrate for α-arabinofuranosidase, an enzyme involved in the breakdown of arabinose-containing polysaccharides. The reagent plays a critical role in studying the enzymatic hydrolysis of arabinofuranoside bonds, particularly in carbohydrate research and the degradation of plant cell wall components. It is widely used in enzymology, glycobiology, and proteomics research to explore the mechanisms of glycoside hydrolases and their role in carbohydrate metabolism.