Learn More
Alkali Scientific p Nitrophenyl a L fucopyranoside, 1 G
SDP
Catalog No. 5000660954
Encompass_Preferred
p-Nitrophenyl α-L-fucopyranoside, available in 1g quantity, is a specialized chemical reagent used in the analysis of carbohydrate structures and enzyme activity. This compound is primarily employed in enzyme assays to study α-L-fucosidases, enzymes responsible for the hydrolysis of fucosidic linkages in glycoproteins. It serves as a substrate that undergoes hydrolysis, producing p-nitrophenol, which can be easily quantified spectrophotometrically. This reagent is crucial for researching metabolic pathways involving fucose, an important sugar in biological processes such as cell recognition and immune response.
Catalog No. 50-006-60954 Supplier Alkali Scientific Supplier No. PHE002CP
p-Nitrophenyl α-L-fucopyranoside, available in 1g quantity, is a specialized chemical reagent used in the analysis of carbohydrate structures and enzyme activity. This compound is primarily employed in enzyme assays to study α-L-fucosidases, enzymes responsible for the hydrolysis of fucosidic linkages in glycoproteins. It serves as a substrate that undergoes hydrolysis, producing p-nitrophenol, which can be easily quantified spectrophotometrically. This reagent is crucial for researching metabolic pathways involving fucose, an important sugar in biological processes such as cell recognition and immune response.