Learn More
Alkali Scientific p Nitrophenyl a L fucopyranoside, 5 G
SDP
Catalog No. 5000661395
Encompass_Preferred
p-Nitrophenyl α-L-fucopyranoside, in a 5g quantity, is a valuable chemical reagent used for enzyme assays in biochemistry and molecular biology. This substrate is commonly used to study α-L-fucosidase activity, which involves the hydrolysis of fucose-containing substrates. Upon enzymatic cleavage, p-nitrophenol is released, allowing for the quantification of enzyme activity through a colorimetric assay. The reagent is highly stable and versatile, making it ideal for use in various research fields, including glycoscience, immunology, and biochemistry, where the investigation of glycosidase enzymes is essential.
Catalog No. 50-006-61395 Supplier Alkali Scientific Supplier No. PHE003CP
p-Nitrophenyl α-L-fucopyranoside, in a 5g quantity, is a valuable chemical reagent used for enzyme assays in biochemistry and molecular biology. This substrate is commonly used to study α-L-fucosidase activity, which involves the hydrolysis of fucose-containing substrates. Upon enzymatic cleavage, p-nitrophenol is released, allowing for the quantification of enzyme activity through a colorimetric assay. The reagent is highly stable and versatile, making it ideal for use in various research fields, including glycoscience, immunology, and biochemistry, where the investigation of glycosidase enzymes is essential.