Learn More
Medchemexpress LLC P-nitrophenyl α-L-fucoside | 10231-84-2 | MFCD00063697 | 98.5% | 285.3 g/mol | C12H15NO7 | 5 MG
SDP
Catalog No. 5000222445
Encompass_Preferred
p-Nitrophenyl α-L-fucoside is a chromogenic substrate used to quantify α-L-fucosidase activity. Enzymatic hydrolysis releases p-nitrophenol, producing a yellow chromophore that can be measured spectrophotometrically for end-point or kinetic assays. Supplied as a high-purity solid for reagent preparation.
  • Chromogenic substrate for α-L-fucosidase assays.
  • Releases p-nitrophenol detectable at ≈405 nm for spectrophotometric readout.
  • High purity (~98.5%) suitable for quantitative enzymatic studies.
  • Supplied as a stable solid for easy dissolution and assay setup.
Catalog No. 50-002-22445 Supplier Medchemexpress LLC Supplier No. HY1344205MG
p-Nitrophenyl α-L-fucoside is a chromogenic substrate used to quantify α-L-fucosidase activity. Enzymatic hydrolysis releases p-nitrophenol, producing a yellow chromophore that can be measured spectrophotometrically for end-point or kinetic assays. Supplied as a high-purity solid for reagent preparation.
  • Chromogenic substrate for α-L-fucosidase assays.
  • Releases p-nitrophenol detectable at ≈405 nm for spectrophotometric readout.
  • High purity (~98.5%) suitable for quantitative enzymatic studies.
  • Supplied as a stable solid for easy dissolution and assay setup.