Learn More
Medchemexpress LLC P-nitrophenyl α-L-fucoside | 10231-84-2 | MFCD00063697 | 98.5% | C12H15NO7 | 10MG
SDP
Catalog No. 5000262008
Encompass_Preferred
p-Nitrophenyl α-L-fucoside is a chromogenic substrate used to determine α-L-fucosidase activity in biochemical assays. Enzymatic hydrolysis releases p-nitrophenol, producing a colorimetric signal measurable by spectrophotometry. The reagent is supplied with high purity for research use.
  • Chromogenic substrate that releases p-nitrophenol upon enzymatic hydrolysis.
  • Enables colorimetric spectrophotometric determination of α-L-fucosidase activity.
  • High purity (98.5%) suitable for assay development and enzyme characterization.
  • Compact milligram package sizes convenient for small-scale experiments.
  • Chemical formula C12H15NO7 and molecular weight 285.25 facilitate calculation of assay concentrations.
Catalog No. 50-002-62008 Supplier Medchemexpress LLC Supplier No. HY13442010MG
p-Nitrophenyl α-L-fucoside is a chromogenic substrate used to determine α-L-fucosidase activity in biochemical assays. Enzymatic hydrolysis releases p-nitrophenol, producing a colorimetric signal measurable by spectrophotometry. The reagent is supplied with high purity for research use.
  • Chromogenic substrate that releases p-nitrophenol upon enzymatic hydrolysis.
  • Enables colorimetric spectrophotometric determination of α-L-fucosidase activity.
  • High purity (98.5%) suitable for assay development and enzyme characterization.
  • Compact milligram package sizes convenient for small-scale experiments.
  • Chemical formula C12H15NO7 and molecular weight 285.25 facilitate calculation of assay concentrations.