Learn More
Alkali Scientific p Nitrophenyl Phosphoryl Choline, 500 Mg
SDP
Catalog No. 5000661191
Encompass_Preferred
p-Nitrophenyl Phosphoryl Choline, 500mg, is a high-quality reagent used for studying enzyme-substrate interactions, particularly in the analysis of phospholipase activity. This choline derivative acts as a substrate for enzymes involved in lipid metabolism, and its hydrolysis releases a p-nitrophenol group, which can be detected spectrophotometrically. The 500mg quantity is ideal for small-scale experimental setups in enzymology, pharmacology, and biochemistry. This reagent is essential in the investigation of phospholipase activities, and its use extends to studies in membrane biology, lipid signaling, and drug development.
Catalog No. 50-006-61191 Supplier Alkali Scientific Supplier No. PNI002CP
p-Nitrophenyl Phosphoryl Choline, 500mg, is a high-quality reagent used for studying enzyme-substrate interactions, particularly in the analysis of phospholipase activity. This choline derivative acts as a substrate for enzymes involved in lipid metabolism, and its hydrolysis releases a p-nitrophenol group, which can be detected spectrophotometrically. The 500mg quantity is ideal for small-scale experimental setups in enzymology, pharmacology, and biochemistry. This reagent is essential in the investigation of phospholipase activities, and its use extends to studies in membrane biology, lipid signaling, and drug development.