Learn More
Medchemexpress LLC p-Nitrophenyl phosphorylcholine | 21064-69-7 | MFCD00077834 | 99.6% | 304.24 | C11H17N2O6P | 10 MG
SDP
Catalog No. 5000215158
Encompass_Preferred
p-Nitrophenyl phosphorylcholine is a chromogenic substrate used to measure phospholipase C (PLC) activity. Upon enzymatic hydrolysis it releases a p-nitrophenolate chromophore that produces a yellow color measurable at 405 nm, enabling straightforward spectrophotometric activity assays.
  • Provides a sensitive colorimetric readout at 405 nm.
  • High chemical purity for reproducible results.
  • Solid, white to off-white appearance for easy handling.
  • Suitable for enzyme activity assays involving phospholipases.
  • Stable under recommended storage conditions for extended shelf life.
Catalog No. 50-002-15158 Supplier Medchemexpress LLC Supplier No. HY10004510MG
p-Nitrophenyl phosphorylcholine is a chromogenic substrate used to measure phospholipase C (PLC) activity. Upon enzymatic hydrolysis it releases a p-nitrophenolate chromophore that produces a yellow color measurable at 405 nm, enabling straightforward spectrophotometric activity assays.
  • Provides a sensitive colorimetric readout at 405 nm.
  • High chemical purity for reproducible results.
  • Solid, white to off-white appearance for easy handling.
  • Suitable for enzyme activity assays involving phospholipases.
  • Stable under recommended storage conditions for extended shelf life.