Learn More
Medchemexpress LLC P-nitrophenyl phosphorylcholine | 21064-69-7 | MFCD00077834 | 99.6% | 304.24 g/mol | C11H17N2O6P | 25 MG
SDP
Catalog No. 5000215655
Encompass_Preferred
p-Nitrophenyl phosphorylcholine is a chromogenic substrate used to measure phospholipase C (PLC) activity. Hydrolysis releases p-nitrophenol, producing a yellow 4-nitrophenolate ion that is typically monitored by absorbance at 405 nm at near-neutral pH. The compound is supplied as a high-purity solid suitable for enzymatic assays and biochemical research.
  • Chromogenic substrate releasing p-nitrophenol measurable at 405 nm.
  • Optimized for PLC activity assays at pH 7.2-7.5.
  • High purity (99.61%) for reliable assay performance.
  • Provided as a solid in small pack sizes starting at 25 mg.
  • Molecular weight 304.24 g/mol for accurate reagent calculations.
Catalog No. 50-002-15655 Supplier Medchemexpress LLC Supplier No. HY10004525MG
p-Nitrophenyl phosphorylcholine is a chromogenic substrate used to measure phospholipase C (PLC) activity. Hydrolysis releases p-nitrophenol, producing a yellow 4-nitrophenolate ion that is typically monitored by absorbance at 405 nm at near-neutral pH. The compound is supplied as a high-purity solid suitable for enzymatic assays and biochemical research.
  • Chromogenic substrate releasing p-nitrophenol measurable at 405 nm.
  • Optimized for PLC activity assays at pH 7.2-7.5.
  • High purity (99.61%) for reliable assay performance.
  • Provided as a solid in small pack sizes starting at 25 mg.
  • Molecular weight 304.24 g/mol for accurate reagent calculations.