Learn More
Medchemexpress LLC P-nitrophenyl phosphorylcholine | 21064-69-7 | MFCD00077834 | 99.6% | 304.24 g/mol | C11H17N2O6P | 5 MG
SDP
Catalog No. 5000215161
Encompass_Preferred
p-Nitrophenyl phosphorylcholine is a chromogenic substrate for measuring phospholipase C (PLC) activity. Enzymatic hydrolysis releases p-nitrophenolate, producing a yellow chromophore that is detectable by spectrophotometry (typically at ~405 nm). The product is supplied with supporting documentation to aid quality control and safe handling.
Catalog No. 50-002-15161 Supplier Medchemexpress LLC Supplier No. HY1000455MG
p-Nitrophenyl phosphorylcholine is a chromogenic substrate for measuring phospholipase C (PLC) activity. Enzymatic hydrolysis releases p-nitrophenolate, producing a yellow chromophore that is detectable by spectrophotometry (typically at ~405 nm). The product is supplied with supporting documentation to aid quality control and safe handling.