Learn More
Medchemexpress LLC P-nitrophenyl phosphorylcholine | 21064-69-7 | MFCD00077834 | 99.6% | 304.24 g·mol⁻¹ | C11H17N2O6P | 50 MG
SDP
Catalog No. 5000215683
Encompass_Preferred
p-Nitrophenyl phosphorylcholine is a chromogenic substrate used to measure phospholipase C (PLC) activity. Hydrolysis by PLC releases p-nitrophenol, which can be quantified by absorbance at 405 nm under near-neutral pH. Supplied as a solid with high reported purity, it is suitable for enzymatic assays, assay development, and kinetic studies.
Catalog No. 50-002-15683 Supplier Medchemexpress LLC Supplier No. HY10004550MG
p-Nitrophenyl phosphorylcholine is a chromogenic substrate used to measure phospholipase C (PLC) activity. Hydrolysis by PLC releases p-nitrophenol, which can be quantified by absorbance at 405 nm under near-neutral pH. Supplied as a solid with high reported purity, it is suitable for enzymatic assays, assay development, and kinetic studies.