Learn More
Cayman Chemical p-Nitrophenyl Sulfate (potassium salt), Purity: ≥98%, CAS: 6217-68-1, 5 g
SDP
Catalog No. 502910554
Encompass_Preferred
A colorimetric arylsulfatase substrate; Upon enzymatic cleavage by arylsulfatase, p-nitrophenyl is released, which can be quantified by colorimetric detection at 400 nm as a measure of arylsulfatase activity
Catalog No. 50-291-0554 Supplier Cayman Chemical Supplier No. 356915 G
A colorimetric arylsulfatase substrate; Upon enzymatic cleavage by arylsulfatase, p-nitrophenyl is released, which can be quantified by colorimetric detection at 400 nm as a measure of arylsulfatase activity