Learn More
Medchemexpress LLC HY-B1986 100mg Medchemexpress, p,p'-DDE CAS:72-55-9 Purity:>98%
SDP
Catalog No. 502031246
Encompass_Preferred
Medchemexpress, HY-B1986 100mg p,p'-DDE CAS:72-55-9 p,p'-DDE (4,4'-DDE), a major metabolite of persistent dichlorodiphenyltrichloroethane (DDT), is a potent androgen receptor antagonist, with an IC50 of 5 μM and a Ki of 3.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1246 Supplier Medchemexpress LLC Supplier No. HYB1986100MG
Medchemexpress, HY-B1986 100mg p,p'-DDE CAS:72-55-9 p,p'-DDE (4,4'-DDE), a major metabolite of persistent dichlorodiphenyltrichloroethane (DDT), is a potent androgen receptor antagonist, with an IC50 of 5 μM and a Ki of 3.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.