Learn More
Medchemexpress LLC p,p'-DDE-d8 | 93952-19-3 | 99% | 5 MG
SDP
Catalog No. 5000411211
Encompass_Preferred
p,p'-DDE-d8 is the deuterium labeled p,p'-DDE. p,p'-DDE (4,4'-DDE), a major metabolite of persistent dichlorodiphenyltrichloroethylene (DDT), is a potent androgen receptor antagonist, with an IC50 of 5 μM and a Ki of 3.5 μM. It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-11211 Supplier Medchemexpress LLC Supplier No. HYB1986S5MG
p,p'-DDE-d8 is the deuterium labeled p,p'-DDE. p,p'-DDE (4,4'-DDE), a major metabolite of persistent dichlorodiphenyltrichloroethylene (DDT), is a potent androgen receptor antagonist, with an IC50 of 5 μM and a Ki of 3.5 μM. It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.