Learn More
Cygnus Technologies P.PASTORIS HCP ELISA KIT,2ND G
Supplier:  Cygnus Technologies F640
Encompass
Our original F140 kit, has a very broad reactivity to peptide epitopes and in addition, also has a small population of antibodies that recognize Pichia glycosylation epitopes. In cases where the recombinant protein product is also glycosylated, there is potential for some cross reactivity that can r
Catalog No. NC0382262