Learn More
ABclonal Technology p-PKC alpha/beta II-T638/T641
SDP
Catalog No. 5000014098
Encompass_Preferred
Protein kinase C (PKC) is a family of serine- and threonine-specific kinases activated by calcium and diacylglycerol PKC family members phosphorylate various protein targets and are involved in multiple cellular signaling pathways They also serve as receptors for phorbol esters tumor-promoting compounds Each PKC member has a distinct expression profile and role in cells The protein encoded by this gene is a PKC family member involved in B cell activation apoptosis endothelial cell proliferation and intestinal sugar absorption Studies in mice suggest it may also regulate neuronal functions and influence fear-induced behavior after stress Alternative splicing generates distinct isoforms of this gene
Catalog No. 50-000-14098 Supplier ABclonal Technology Supplier No. AP137520UL
Protein kinase C (PKC) is a family of serine- and threonine-specific kinases activated by calcium and diacylglycerol PKC family members phosphorylate various protein targets and are involved in multiple cellular signaling pathways They also serve as receptors for phorbol esters tumor-promoting compounds Each PKC member has a distinct expression profile and role in cells The protein encoded by this gene is a PKC family member involved in B cell activation apoptosis endothelial cell proliferation and intestinal sugar absorption Studies in mice suggest it may also regulate neuronal functions and influence fear-induced behavior after stress Alternative splicing generates distinct isoforms of this gene