Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Propoxyaniline, MP Biomedicals

Catalog No. ICN217491 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN217491 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN217491 Supplier MP Biomedicals, Inc Supplier No. 0521749101

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4469-80-1
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00043621
InChI Key DWOIGSLSPPLRKO-UHFFFAOYSA-N
Synonym p-propoxyaniline, benzenamine, 4-propoxy, 4-n-propoxyaniline, 4-propoxyphenyl amine hydrochloride, 4-propoxyphenylamine, hydrochloride, 4-propyloxyaniline, 4-propoxybenzenamine, 4-propoxy-phenylamine
PubChem CID 78221
IUPAC Name 4-propoxyaniline
SMILES CCCOC1=CC=C(N)C=C1

Specifications

Quantity 10 g
Chemical Name or Material p-Propoxyaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.