Learn More
SPEX CERTIPREP LLC 3P Quantitative Enterococcus/Fecal Strep
Catalog No. NC0812602
Encompass_Preferred
(NSI by Zeptometrix) Designed for use with all MPN and MF procedures. Derived from E. faecalis (NCTC 775) and preserved in our proprietary Microgel-Flash quick dissolve matrix in the range of 20 - 1000 CFU/MPN per 100 mL. Sample taken from previous Interlab PT study. Accompanying COA includes study statistics. Sterile hydration buffer included. Store at -10C to -20C.
Catalog No. NC0812602 Supplier SPEX CERTIPREP LLC 3P Supplier No. MICQC5
(NSI by Zeptometrix) Designed for use with all MPN and MF procedures. Derived from E. faecalis (NCTC 775) and preserved in our proprietary Microgel-Flash quick dissolve matrix in the range of 20 - 1000 CFU/MPN per 100 mL. Sample taken from previous Interlab PT study. Accompanying COA includes study statistics. Sterile hydration buffer included. Store at -10C to -20C.