Learn More
Medchemexpress LLC p-SCN-Bn-deferoxamine | 1222468-90-7 | 98.0% | C33H52N8O8S2 | 10MG
SDP
Catalog No. 5000259253
Encompass_Preferred
p-SCN-Bn-deferoxamine is a bifunctional chelator bearing a para-isothiocyanate benzyl reactive group used to attach chelators to amine-containing biomolecules such as antibodies, enabling radiolabeling and bioconjugation workflows. Supplied as a white to off-white solid with a reported purity of 98.0%, it is intended for research use.
  • Provides an isothiocyanate reactive handle for stable attachment to amines.
  • Enables chelator-based radiolabeling and antibody conjugation workflows.
  • High purity solid suitable for research applications.
  • Soluble in DMSO (≈70 mg/mL with sonication and warming).
  • Available in small research sizes for development and optimization.
Catalog No. 50-002-59253 Supplier Medchemexpress LLC Supplier No. HY13479710MG
p-SCN-Bn-deferoxamine is a bifunctional chelator bearing a para-isothiocyanate benzyl reactive group used to attach chelators to amine-containing biomolecules such as antibodies, enabling radiolabeling and bioconjugation workflows. Supplied as a white to off-white solid with a reported purity of 98.0%, it is intended for research use.
  • Provides an isothiocyanate reactive handle for stable attachment to amines.
  • Enables chelator-based radiolabeling and antibody conjugation workflows.
  • High purity solid suitable for research applications.
  • Soluble in DMSO (≈70 mg/mL with sonication and warming).
  • Available in small research sizes for development and optimization.