Learn More
Medchemexpress LLC p-SCN-Bn-DOTA 100mg | 127985-74-4 | 100 MG
SDP
Catalog No. 5000307752
Encompass_Preferred
p-SCN-Bn-DOTA is a bifunctional chelating agent that couples a reactive para-isothiocyanatobenzyl group to a DOTA macrocycle, enabling covalent conjugation to biomolecules and stable chelation of radiometals for radiolabeling and radioimmunoconjugation applications.
  • Reactive para-isothiocyanatobenzyl group for covalent attachment to amine groups on proteins.
  • DOTA macrocycle provides stable complexation with a range of radiometals.
  • High purity suitable for research and analytical use (99.8%).
  • Available as a solid and as 10 mM solutions in DMSO for convenient handling.
  • Intended for radiolabeling, radioimmunoconjugation, and preclinical imaging studies.
Catalog No. 50-003-07752 Supplier Medchemexpress LLC Supplier No. HY114377100MG
p-SCN-Bn-DOTA is a bifunctional chelating agent that couples a reactive para-isothiocyanatobenzyl group to a DOTA macrocycle, enabling covalent conjugation to biomolecules and stable chelation of radiometals for radiolabeling and radioimmunoconjugation applications.
  • Reactive para-isothiocyanatobenzyl group for covalent attachment to amine groups on proteins.
  • DOTA macrocycle provides stable complexation with a range of radiometals.
  • High purity suitable for research and analytical use (99.8%).
  • Available as a solid and as 10 mM solutions in DMSO for convenient handling.
  • Intended for radiolabeling, radioimmunoconjugation, and preclinical imaging studies.