Learn More
Medchemexpress LLC p-SCN-Bn-DOTA 10mM 1mL | 127985-74-4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11437710MM 1ML
Encompass_Preferred
p-SCN-Bn-DOTA is a bifunctional chelating agent with a p-isothiocyanatobenzyl reactive group for covalent attachment to biomolecules and a DOTA macrocycle for stable radiometal chelation. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for research use in antibody labeling, radiolabeling, and imaging studies.
  • Bifunctional chelator enabling covalent conjugation and metal complexation.
  • P-isothiocyanatobenzyl group for stable attachment to primary amines.
  • DOTA macrocycle forms high-stability complexes with radiometals.
  • Supplied as 10 mM solution in DMSO, ready for conjugation.
  • High purity suitable for research applications.
  • Intended for antibody labeling, imaging, and radiotherapeutic research.
Catalog No. 50-002-28870