Learn More
Medchemexpress LLC p-SCN-Bn-DOTA 25mg | 127985-74-4 | 25 MG
SDP
Catalog No. 5000307584
Encompass_Preferred
p-SCN-Bn-DOTA is a bifunctional chelating agent that combines an isothiocyanate reactive group for covalent protein conjugation with a DOTA macrocycle for stable radiometal chelation. It is used to conjugate radionuclides to antibodies and other biomolecules for radioimaging and radioimmunotherapy applications.
  • Bifunctional chelator with an isothiocyanate reactive group for protein conjugation.
  • DOTA macrocycle enables stable chelation of radiometals.
  • Suitable for antibody conjugation and radioimmunoassays.
  • Available as a solid powder, typically supplied in milligram-scale quantities.
  • High purity suitable for research applications.
Catalog No. 50-003-07584 Supplier Medchemexpress LLC Supplier No. HY11437725MG
p-SCN-Bn-DOTA is a bifunctional chelating agent that combines an isothiocyanate reactive group for covalent protein conjugation with a DOTA macrocycle for stable radiometal chelation. It is used to conjugate radionuclides to antibodies and other biomolecules for radioimaging and radioimmunotherapy applications.
  • Bifunctional chelator with an isothiocyanate reactive group for protein conjugation.
  • DOTA macrocycle enables stable chelation of radiometals.
  • Suitable for antibody conjugation and radioimmunoassays.
  • Available as a solid powder, typically supplied in milligram-scale quantities.
  • High purity suitable for research applications.