Learn More
Medchemexpress LLC p-SCN-Bn-DOTA 50mg | 127985-74-4 | 50 MG
SDP
Catalog No. 5000228869
Encompass_Preferred
p-SCN-Bn-DOTA is a bifunctional chelating reagent that couples a para-isothiocyanatobenzyl reactive group to a DOTA macrocycle, enabling covalent conjugation to biomolecules and stable chelation of radiometals for radioimmunoconjugation and radiopharmaceutical research.
  • Enables covalent attachment to proteins and antibodies via an isothiocyanate group.
  • Provides stable complexation of radiometals through the DOTA chelator.
  • High purity suitable for research-grade radioconjugation.
  • Solid, easy-to-weigh form for small-scale conjugation workflows.
  • Manufacturer provides SDS and datasheet for safe handling and use.
Catalog No. 50-002-28869 Supplier Medchemexpress LLC Supplier No. HY11437750MG
p-SCN-Bn-DOTA is a bifunctional chelating reagent that couples a para-isothiocyanatobenzyl reactive group to a DOTA macrocycle, enabling covalent conjugation to biomolecules and stable chelation of radiometals for radioimmunoconjugation and radiopharmaceutical research.
  • Enables covalent attachment to proteins and antibodies via an isothiocyanate group.
  • Provides stable complexation of radiometals through the DOTA chelator.
  • High purity suitable for research-grade radioconjugation.
  • Solid, easy-to-weigh form for small-scale conjugation workflows.
  • Manufacturer provides SDS and datasheet for safe handling and use.