Learn More
Medchemexpress LLC P-SCN-Bn-TCMC | 282097-63-6 | 547.67 | 25 MG
SDP
Catalog No. 5000403370
Encompass_Preferred
p-SCN-Bn-TCMC is a bifunctional chelator. Its cyclen backbone binds Hg2+, while its isothiocyanate group allows for conjugation to biomolecules. p-SCN-Bn-TCMC can be used in the research of prostate cancer.
  • Bifunctional chelator
  • Cyclen backbone binds Hg2+
  • Isothiocyanate group allows for conjugation to biomolecules
  • Used in the research of prostate cancer
Catalog No. 50-004-03370 Supplier Medchemexpress LLC Supplier No. 25MGHY201263
p-SCN-Bn-TCMC is a bifunctional chelator. Its cyclen backbone binds Hg2+, while its isothiocyanate group allows for conjugation to biomolecules. p-SCN-Bn-TCMC can be used in the research of prostate cancer.
  • Bifunctional chelator
  • Cyclen backbone binds Hg2+
  • Isothiocyanate group allows for conjugation to biomolecules
  • Used in the research of prostate cancer