Learn More
Creative Biomart P-Selectin Glycoprotein Ligand-1 (PSGL-1)
Supplier:  Creative Biomart THP0272
Encompass_Preferred
Recombinant Human P-Selectin Glycoprotein Ligand-1 (PSGL-1) Protein is produced by CHO expression system. N-terminal Sequence Analysis: Gln42.
Catalog No. 50-208-4323