Learn More
Medchemexpress LLC P-Selectin Human B 20ug | 20ug
SDP
Catalog No. 5000193797
Encompass_Preferred
P-selectin is a Ca(2 )-dependent receptor on myeloid cells that critically binds to sialic acid-Lewis X on neutrophils and monocytes promoting the interaction between activated endothelial cells or platelets and leukocytes interaction This binding primarily to SELPLG/PSGL1 and PODXL2 is critical for rapid rolling of leukocytes during early inflammation P-Selectin Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived P-Selectin protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-93797 Supplier Medchemexpress LLC Supplier No. HYP7820120UG
P-selectin is a Ca(2 )-dependent receptor on myeloid cells that critically binds to sialic acid-Lewis X on neutrophils and monocytes promoting the interaction between activated endothelial cells or platelets and leukocytes interaction This binding primarily to SELPLG/PSGL1 and PODXL2 is critical for rapid rolling of leukocytes during early inflammation P-Selectin Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived P-Selectin protein expressed by HEK293 with C-Avi C-His labeled tag