Learn More
Medchemexpress LLC P-Selectin Human H 10ug
SDP
Catalog No. 5000179562
Encompass_Preferred
P-selectin is a Ca(2 )-dependent receptor on myeloid cells that critically binds to sialic acid-Lewis X on neutrophils and monocytes promoting the interaction between activated endothelial cells or platelets and leukocytes interaction This binding primarily to SELPLG/PSGL1 and PODXL2 is critical for rapid rolling of leukocytes during early inflammation P-Selectin Protein Human (HEK293 Fc) is the recombinant human-derived P-Selectin protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-79562 Supplier Medchemexpress LLC Supplier No. HYP7369010UG
P-selectin is a Ca(2 )-dependent receptor on myeloid cells that critically binds to sialic acid-Lewis X on neutrophils and monocytes promoting the interaction between activated endothelial cells or platelets and leukocytes interaction This binding primarily to SELPLG/PSGL1 and PODXL2 is critical for rapid rolling of leukocytes during early inflammation P-Selectin Protein Human (HEK293 Fc) is the recombinant human-derived P-Selectin protein expressed by HEK293 with C-hFc labeled tag