Learn More
Medchemexpress LLC P-selectin Mouse B 100ug
SDP
Catalog No. 5000181936
Encompass_Preferred
P-selectin is a Ca(2 )-dependent receptor on myeloid cells that specifically binds to carbohydrates on neutrophils and monocytes It mediates interactions between activated endothelial cells or platelets and leukocytes by recognizing sialic acid-Lewis X P-selectin Protein Mouse (Biotinylated HEK293 Avi-His) is the recombinant mouse-derived P-selectin protein expressed by HEK293 with C-Avi C-6 His labeled tag
Catalog No. 50-001-81936 Supplier Medchemexpress LLC Supplier No. HYP72408100UG
P-selectin is a Ca(2 )-dependent receptor on myeloid cells that specifically binds to carbohydrates on neutrophils and monocytes It mediates interactions between activated endothelial cells or platelets and leukocytes by recognizing sialic acid-Lewis X P-selectin Protein Mouse (Biotinylated HEK293 Avi-His) is the recombinant mouse-derived P-selectin protein expressed by HEK293 with C-Avi C-6 His labeled tag