Learn More
Medchemexpress LLC P-Selectin Mouse H 50ug | 50ug
SDP
Catalog No. 5000193649
Encompass_Preferred
P-selectin is a Ca(2 )-dependent receptor on myeloid cells that specifically binds to carbohydrates on neutrophils and monocytes It mediates interactions between activated endothelial cells or platelets and leukocytes by recognizing sialic acid-Lewis X P-Selectin Protein Mouse (HEK293 His) is the recombinant mouse-derived P-Selectin protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-93649 Supplier Medchemexpress LLC Supplier No. HYP7834950UG
P-selectin is a Ca(2 )-dependent receptor on myeloid cells that specifically binds to carbohydrates on neutrophils and monocytes It mediates interactions between activated endothelial cells or platelets and leukocytes by recognizing sialic acid-Lewis X P-Selectin Protein Mouse (HEK293 His) is the recombinant mouse-derived P-Selectin protein expressed by HEK293 with C-His labeled tag