Learn More
Medchemexpress LLC P-Selectin Rat HEK 100ug
SDP
Catalog No. 5000179645
Encompass_Preferred
P-Selectin a Ca(2 )-dependent receptor on myeloid cells binds to neutrophils and monocytes via carbohydrates It interacts with SELPLG to enable rapid leukocyte rolling over vascular surfaces in early inflammation P-Selectin also interacts with SNX17 and promotes neutrophil adhesion and rolling requiring SELPLG s sialyl-Lewis X and tyrosine sulfation P-Selectin Protein Rat (HEK293 His) is the recombinant rat-derived P-Selectin protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79645 Supplier Medchemexpress LLC Supplier No. HYP73689100UG
P-Selectin a Ca(2 )-dependent receptor on myeloid cells binds to neutrophils and monocytes via carbohydrates It interacts with SELPLG to enable rapid leukocyte rolling over vascular surfaces in early inflammation P-Selectin also interacts with SNX17 and promotes neutrophil adhesion and rolling requiring SELPLG s sialyl-Lewis X and tyrosine sulfation P-Selectin Protein Rat (HEK293 His) is the recombinant rat-derived P-Selectin protein expressed by HEK293 with C-His labeled tag