Learn More
Medchemexpress LLC P-Selectin Rhesus M 10ug
SDP
Catalog No. 5000179559
Encompass_Preferred
P-Selectin Protein Rhesus Macaque (HEK293 His) is a member of the selectin family of cell adhesion molecules P-selectin (CD62P) has an N-terminal lectin domain an epidermal growth factor motif (generally) nine regulatory protein repeats a transmembrane section and a short intracytoplasmic tail P-selectin mediates leukocyte rolling on stimulated endothelial cells and heterotypic aggregation of activated platelets onto leukocytes P-selectin mediates heterotypic aggregation of activated platelets to cancer cells and adhesion of cancer cells to stimulated endothelial cells P-selectin glycoprotein ligand-1 (PSGL-1) is a major ligand for P-selectin[1][2][3] P-Selectin Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived P-Selectin protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79559 Supplier Medchemexpress LLC Supplier No. HYP7368710UG
P-Selectin Protein Rhesus Macaque (HEK293 His) is a member of the selectin family of cell adhesion molecules P-selectin (CD62P) has an N-terminal lectin domain an epidermal growth factor motif (generally) nine regulatory protein repeats a transmembrane section and a short intracytoplasmic tail P-selectin mediates leukocyte rolling on stimulated endothelial cells and heterotypic aggregation of activated platelets onto leukocytes P-selectin mediates heterotypic aggregation of activated platelets to cancer cells and adhesion of cancer cells to stimulated endothelial cells P-selectin glycoprotein ligand-1 (PSGL-1) is a major ligand for P-selectin[1][2][3] P-Selectin Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived P-Selectin protein expressed by HEK293 with C-His labeled tag