Learn More
Cayman Chemical P-SelectInCD62Phumnreco 100ug
SDP
Catalog No. 502913627
Encompass_Preferred
Source: Active recombinant C-terminal human IgG1 Fc-tagged P-selectin expressed in HEK293 cells • Amino acids: 42-771 • MW: 107 kDa
Catalog No. 50-291-3627 Supplier Cayman Chemical Supplier No. 35508100 UG
Source: Active recombinant C-terminal human IgG1 Fc-tagged P-selectin expressed in HEK293 cells • Amino acids: 42-771 • MW: 107 kDa