Learn More
Cayman Chemical P-SelectInCD62Pmousereco 100ug
SDP
Catalog No. 502910204
Encompass_Preferred
Source: Active recombinant C-terminal His-tagged P-selectin expressed in HEK293 cells • Amino acids: 42-709 • MW: 74 kDa
Catalog No. 50-291-0204 Supplier Cayman Chemical Supplier No. 35728100 UG
Source: Active recombinant C-terminal His-tagged P-selectin expressed in HEK293 cells • Amino acids: 42-709 • MW: 74 kDa